Assay Details
| Gene Symbol: jag1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131861.1 | NP_571936.1 | 25 - 26 | 3258 | 107 | |
| XM_005168204.4 | XP_005168261.1 | 24 - 25 | 3335 | 107 | ||
| XM_005168205.4 | XP_005168262.1 | 24 - 25 | 3335 | 107 | ||
| GenBank mRNA | AF229448.1 | - | 25 - 26 | 3258 | 107 | |
| AY221107.1 | - | 25 - 26 | 3192 | 107 | ||
| BC162468.1 | - | 25 - 26 | 107 | |||
More Information
Set Membership: |
|

jag1a
Genomic Map