Assay Details
| Gene Symbol: psap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309267.1 | NP_001296196.1 | 11 - 12 | 1441 | 81 | |
| NM_131883.2 | NP_571958.2 | 12 - 13 | 1447 | 81 | ||
| GenBank mRNA | AF108655.1 | - | 12 - 13 | 1426 | 81 | |
| AF276996.1 | - | 11 - 12 | 1404 | 81 | ||
| BC056717.1 | - | 11 - 12 | 1410 | 81 | ||
| BC063994.1 | - | 11 - 12 | 1408 | 81 | ||
| DT076525.1 | - | 4 - 5 | 466 | 81 | ||
More Information
Set Membership: |
|

psap
Genomic Map