Assay Details
Gene Symbol: p2rx4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153653.1 | NP_705939.1 | 2 - 3 | 315 | 71 | |
XM_021468749.1 | XP_021324424.1 | 2 - 3 | 454 | 71 | ||
GenBank mRNA | AF317643.1 | - | 2 - 3 | 315 | 71 | |
BC066495.1 | - | 2 - 3 | 304 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
