Assay Details
| Gene Symbol: sec61a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153659.1 | NP_705945.1 | 9 - 10 | 1104 | 85 | |
| GenBank mRNA | AY029527.1 | - | 9 - 10 | 1104 | 85 | |
| BC044351.1 | - | 9 - 10 | 1086 | 85 | ||
| BC066715.1 | - | 9 - 10 | 85 | |||
More Information
Set Membership: |
|

sec61a1
Genomic Map