Assay Details
| Gene Symbol: ckbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173222.1 | NP_775329.1 | 4 - 5 | 559 | 77 | |
| GenBank mRNA | AY055849.1 | - | 4 - 5 | 559 | 77 | |
| BC049529.1 | - | 4 - 5 | 575 | 77 | ||
| BC064285.1 | - | 4 - 5 | 563 | 77 | ||
More Information
Set Membership: |
|

ckbb
Genomic Map