Assay Details
Gene Symbol: taf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173260.1 | NP_775367.1 | 8 - 9 | 687 | 100 | |
XM_005173300.4 | XP_005173357.1 | 8 - 9 | 851 | 100 | ||
GenBank mRNA | AF506232.1 | - | 8 - 9 | 687 | 100 | |
BC071324.1 | - | 8 - 9 | 651 | 100 | ||
BC154771.1 | - | 8 - 9 | 821 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
