Assay Details
| Gene Symbol: opn1mw2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182891.2 | NP_878311.1 | 2 - 3 | 575 | 89 | |
| GenBank mRNA | BC055605.1 | - | 2 - 3 | 575 | 89 | |
| BC164163.1 | - | 2 - 3 | 561 | 89 | ||
| KT008395.1 | - | 2 - 3 | 527 | 89 | ||
More Information
Set Membership: |
|

opn1mw2
Genomic Map