Assay Details
| Gene Symbol: gatm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199531.1 | NP_955825.1 | 5 - 6 | 862 | 61 | |
| GenBank mRNA | BC056747.1 | - | 5 - 6 | 862 | 61 | |
| BC067594.1 | - | 5 - 6 | 862 | 61 | ||
More Information
Set Membership: |
|

gatm
Genomic Map