Assay Details
| Gene Symbol: polr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199669.1 | NP_955963.1 | 3 - 4 | 393 | 133 | |
| GenBank mRNA | BC045299.1 | - | 3 - 4 | 393 | 133 | |
| BC164883.1 | - | 2 - 3 | 315 | 133 | ||
More Information
Set Membership: |
|

polr2g
Genomic Map