Assay Details
Gene Symbol: dtnba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001324537.1 | NP_001311466.1 | 14 - 15 | 1666 | 103 | |
NM_199709.2 | NP_956003.2 | 13 - 14 | 1581 | 103 | ||
XM_005168526.4 | XP_005168583.1 | 15 - 16 | 1711 | 103 | ||
XM_005168527.4 | XP_005168584.1 | 15 - 16 | 1711 | 103 | ||
XM_005168528.4 | XP_005168585.1 | 15 - 16 | 1711 | 103 | ||
XM_009294981.3 | XP_009293256.1 | 14 - 15 | 1668 | 103 | ||
GenBank mRNA | AY423026.1 | - | 13 - 14 | 1382 | 103 | |
DQ516346.1 | - | 14 - 15 | 1466 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
