Assay Details
Gene Symbol: dtnba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001324537.1 | NP_001311466.1 | 7 - 8 | 940 | 62 | |
NM_199709.2 | NP_956003.2 | 6 - 7 | 855 | 62 | ||
XM_005168526.4 | XP_005168583.1 | 8 - 9 | 985 | 62 | ||
XM_005168527.4 | XP_005168584.1 | 8 - 9 | 985 | 62 | ||
XM_005168528.4 | XP_005168585.1 | 8 - 9 | 985 | 62 | ||
XM_009294981.3 | XP_009293256.1 | 7 - 8 | 942 | 62 | ||
GenBank mRNA | AY423026.1 | - | 6 - 7 | 656 | 62 | |
BC078203.1 | - | 8 - 9 | 938 | 62 | ||
DQ516346.1 | - | 7 - 8 | 740 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
