Assay Details
| Gene Symbol: mob3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199716.1 | NP_956010.1 | 3 - 4 | 916 | 120 | |
| GenBank mRNA | BC049527.1 | - | 3 - 4 | 916 | 120 | |
| BC067183.1 | - | 3 - 4 | 750 | 120 | ||
More Information
Set Membership: |
|

mob3a
Genomic Map