Assay Details
| Gene Symbol: adsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199899.2 | NP_956193.2 | 10 - 11 | 1259 | 67 | |
| GenBank mRNA | AY398306.1 | - | 10 - 11 | 1256 | 67 | |
| BC045891.1 | - | 10 - 11 | 1154 | 67 | ||
| BC071343.1 | - | 10 - 11 | 1125 | 67 | ||
| BC154295.1 | - | 10 - 11 | 1131 | 67 | ||
| BC164214.1 | - | 10 - 11 | 1122 | 67 | ||
| EH571395.1 | - | 3 - 4 | 425 | 67 | ||
| EH606697.1 | - | 3 - 4 | 434 | 67 | ||
More Information
Set Membership: |
|

adsl
Genomic Map