Assay Details
| Gene Symbol: nme2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199970.2 | 2 - 3 | 214 | 88 | ||
| XM_021470954.1 | XP_021326629.1 | 2 - 3 | 296 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nme2a| Gene Symbol: nme2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199970.2 | 2 - 3 | 214 | 88 | ||
| XM_021470954.1 | XP_021326629.1 | 2 - 3 | 296 | 88 | ||
Set Membership: |
|