Assay Details
| Gene Symbol: ctss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200426.1 | NP_956720.1 | 4 - 5 | 422 | 81 | |
| XM_017352203.2 | XP_017207692.1 | 3 - 4 | 462 | 81 | ||
| XM_017352204.2 | XP_017207693.1 | 4 - 5 | 885 | 81 | ||
| GenBank mRNA | BC054668.1 | - | 4 - 5 | 422 | 81 | |
| BC066625.1 | - | 3 - 4 | 374 | 81 | ||
More Information
Set Membership: |
|

ctss1
Genomic Map