Assay Details
Gene Symbol: acsl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200649.1 | NP_956943.1 | 9 - 10 | 1460 | 98 | |
GenBank mRNA | BC057476.1 | - | 9 - 10 | 1460 | 98 | |
BC164569.1 | - | 8 - 9 | 1284 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
