Assay Details
| Gene Symbol: ndufa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200714.1 | NP_957008.1 | 188 | 58 | ||
| GenBank mRNA | BC059474.1 | - | 188 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ndufa13| Gene Symbol: ndufa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200714.1 | NP_957008.1 | 188 | 58 | ||
| GenBank mRNA | BC059474.1 | - | 188 | 58 | ||
Set Membership: |
|