Assay Details
| Gene Symbol: nudt21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201117.1 | NP_957411.1 | 1 - 2 | 252 | 63 | |
| GenBank mRNA | BC053172.1 | - | 1 - 2 | 252 | 63 | |
| BC165427.1 | - | 1 - 2 | 159 | 63 | ||
More Information
Set Membership: |
|

nudt21
Genomic Map