Assay Details
| Gene Symbol: eif4a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201510.1 | NP_958918.1 | 10 - 11 | 1100 | 64 | |
| GenBank mRNA | BC049427.1 | - | 10 - 11 | 1100 | 64 | |
| BC153507.1 | - | 1421 | 64 | |||
More Information
Set Membership: |
|

eif4a1b
Genomic Map