Assay Details
| Gene Symbol: bcl7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212560.1 | NP_997725.1 | 2 - 3 | 236 | 97 | |
| XM_005165362.4 | XP_005165419.1 | 2 - 3 | 300 | 97 | ||
| XM_021476311.1 | XP_021331986.1 | 2 - 3 | 98 | 97 | ||
| GenBank mRNA | AY394963.1 | - | 2 - 3 | 236 | 97 | |
More Information
Set Membership: |
|

bcl7a
Genomic Map