Assay Details
| Gene Symbol: atp5pf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213307.1 | NP_998472.1 | 2 - 3 | 220 | 93 | |
| XM_005167990.4 | XP_005168047.1 | 2 - 3 | 300 | 93 | ||
| XM_017352970.2 | XP_017208459.1 | 2 - 3 | 286 | 93 | ||
| XM_017352984.2 | XP_017208473.1 | 2 - 3 | 283 | 93 | ||
| GenBank mRNA | BC066613.1 | - | 2 - 3 | 220 | 93 | |
More Information
Set Membership: |
|

atp5pf
Genomic Map