Assay Details
| Gene Symbol: pla2g12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213430.1 | NP_998595.2 | 1 - 2 | 400 | 123 | |
| GenBank mRNA | BC052113.1 | - | 1 - 2 | 400 | 123 | |
| BC065857.1 | - | 1 - 2 | 378 | 123 | ||
| BC155273.1 | - | 1 - 2 | 406 | 123 | ||
More Information
Set Membership: |
|

pla2g12b
Genomic Map