Assay Details
| Gene Symbol: tsg101a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002089.1 | NP_001002089.1 | 9 - 10 | 1372 | 72 | |
| GenBank mRNA | BC071540.1 | - | 9 - 10 | 1372 | 72 | |
More Information
Set Membership: |
|

tsg101a
Genomic Map