Assay Details
Gene Symbol: ptges3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002137.2 | NP_001002137.1 | 6 - 7 | 532 | 99 | |
GenBank mRNA | BC071423.1 | - | 6 - 7 | 534 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
