Assay Details
Gene Symbol: atp6v1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002526.2 | NP_001002526.1 | 1 - 2 | 261 | 81 | |
GenBank mRNA | AY648759.1 | - | 1 - 2 | 222 | 81 | |
BC076373.1 | - | 1 - 2 | 236 | 81 | ||
BC164921.1 | - | 1 - 2 | 191 | 81 | ||
EH549763.1 | - | 1 - 2 | 302 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
