Assay Details
| Gene Symbol: ndufv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003747.1 | NP_001003747.1 | 9 - 10 | 1283 | 58 | |
| GenBank mRNA | BC078648.1 | - | 9 - 10 | 1283 | 58 | |
More Information
Set Membership: |
|

ndufv1
Genomic Map