Assay Details
| Gene Symbol: snrnp70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003875.2 | NP_001003875.1 | 5 - 6 | 543 | 71 | |
| XM_017354005.2 | XP_017209494.1 | 5 - 6 | 540 | 71 | ||
| GenBank mRNA | AY648811.1 | - | 5 - 6 | 488 | 71 | |
| BC077089.1 | - | 5 - 6 | 499 | 71 | ||
| BC095150.1 | - | 5 - 6 | 518 | 71 | ||
| BC117602.1 | - | 5 - 6 | 530 | 71 | ||
More Information
Set Membership: |
|

snrnp70
Genomic Map