Assay Details
| Gene Symbol: ccnd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006017.1 | NP_001006017.1 | 4 - 5 | 898 | 67 | |
| GenBank mRNA | BC083406.1 | - | 4 - 5 | 898 | 67 | |
| BC163994.1 | - | 4 - 5 | 777 | 67 | ||
More Information
Set Membership: |
|

ccnd3
Genomic Map