Assay Details
| Gene Symbol: nudt3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006034.2 | NP_001006034.1 | 4 - 5 | 502 | 84 | |
| GenBank mRNA | BC083382.1 | - | 4 - 5 | 503 | 84 | |
More Information
Set Membership: |
|

nudt3b
Genomic Map