Assay Details
| Gene Symbol: ralab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007321.1 | NP_001007322.1 | 3 - 4 | 489 | 87 | |
| GenBank mRNA | BC085637.1 | - | 3 - 4 | 489 | 87 | |
More Information
Set Membership: |
|

ralab
Genomic Map