Assay Details
| Gene Symbol: cetp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007360.1 | NP_001007361.1 | 2 - 3 | 258 | 77 | |
| GenBank mRNA | BC085584.1 | - | 2 - 3 | 258 | 77 | |
More Information
Set Membership: |
|

cetp
Genomic Map