Assay Details
| Gene Symbol: cetp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007360.1 | NP_001007361.1 | 8 - 9 | 784 | 106 | |
| XM_009293552.3 | XP_009291827.1 | 7 - 8 | 1007 | 106 | ||
| GenBank mRNA | BC085584.1 | - | 8 - 9 | 784 | 106 | |
More Information
Set Membership: |
|

cetp
Genomic Map