Assay Details
| Gene Symbol: bfsp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008633.2 | NP_001008633.1 | 1 - 2 | 524 | 65 | |
| XM_005163493.4 | XP_005163550.1 | 1 - 2 | 935 | 65 | ||
| XM_009298615.3 | XP_009296890.1 | 1 - 2 | 935 | 65 | ||
| GenBank mRNA | BC086849.1 | - | 1 - 2 | 526 | 65 | |
| BC165202.1 | - | 1 - 2 | 462 | 65 | ||
More Information
Set Membership: |
|

bfsp2
Genomic Map