Assay Details
| Gene Symbol: fgfrl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012263.2 | NP_001012263.2 | 1 - 2 | 93 | 78 | |
| XM_005173306.4 | XP_005173363.1 | 2 - 3 | 339 | 78 | ||
| GenBank mRNA | AJ781308.2 | - | 1 - 2 | 93 | 78 | |
| AJ781309.1 | - | 1 - 2 | 93 | 78 | ||
| BC162498.1 | - | 1 - 2 | 93 | 78 | ||
| CU458876.1 | - | 1 - 2 | 67 | 78 | ||
More Information
Set Membership: |
|

fgfrl1b
Genomic Map