Assay Details
| Gene Symbol: slc25a44b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013519.1 | NP_001013537.1 | 2 - 3 | 798 | 66 | |
| GenBank mRNA | BC091444.1 | - | 2 - 3 | 798 | 66 | |
More Information
Set Membership: |
|

slc25a44b
Genomic Map