Assay Details
| Gene Symbol: asrgl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013529.1 | NP_001013547.1 | 3 - 4 | 428 | 61 | |
| GenBank mRNA | BC090901.1 | - | 3 - 4 | 428 | 61 | |
| BC164091.1 | - | 2 - 3 | 361 | 61 | ||
More Information
Set Membership: |
|

asrgl1
Genomic Map