Assay Details
| Gene Symbol: sept9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014307.3 | NP_001014329.1 | 8 - 9 | 928 | 72 | |
| NM_001327974.1 | NP_001314903.1 | 10 - 11 | 1660 | 72 | ||
| NM_001327975.1 | NP_001314904.1 | 10 - 11 | 1560 | 72 | ||
| NM_001327977.1 | NP_001314906.1 | 8 - 9 | 916 | 72 | ||
| GenBank mRNA | BC091861.1 | - | 8 - 9 | 932 | 72 | |
| BC164371.1 | - | 7 - 8 | 836 | 72 | ||
| HM208344.1 | - | 8 - 9 | 925 | 72 | ||
| HM208345.1 | - | 8 - 9 | 916 | 72 | ||
More Information
Set Membership: |
|

sept9b
Genomic Map