Assay Details
Gene Symbol: med10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017699.1 | NP_001017699.1 | 2 - 3 | 323 | 78 | |
GenBank mRNA | BC093274.1 | - | 2 - 3 | 323 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
