Assay Details
| Gene Symbol: arl4ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017749.1 | NP_001017749.1 | 1 | 250 | 82 | |
| GenBank mRNA | BC093202.1 | - | 1 | 250 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

arl4ca| Gene Symbol: arl4ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017749.1 | NP_001017749.1 | 1 | 250 | 82 | |
| GenBank mRNA | BC093202.1 | - | 1 | 250 | 82 | |
Set Membership: |
|