Assay Details
| Gene Symbol: dnmt3aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018134.1 | NP_001018144.1 | 6 - 7 | 864 | 65 | |
| XM_005160858.4 | XP_005160915.1 | 9 - 10 | 1759 | 65 | ||
| GenBank mRNA | AB196919.1 | - | 6 - 7 | 864 | 65 | |
| BC162629.1 | - | 5 - 6 | 861 | 65 | ||
More Information
Set Membership: |
|

dnmt3aa
Genomic Map