Assay Details
| Gene Symbol: lrit1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018164.2 | NP_001018174.2 | 3 - 4 | 1187 | 77 | |
| GenBank mRNA | BC095674.1 | - | 3 - 4 | 1004 | 77 | |
| CU468741.1 | - | 3 - 4 | 887 | 77 | ||
More Information
Set Membership: |
|

lrit1a
Genomic Map