Assay Details
Gene Symbol: sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024377.2 | NP_001019548.1 | 3 - 4 | 351 | 83 | |
GenBank mRNA | BC059552.1 | - | 3 - 4 | 281 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
