Assay Details
Gene Symbol: tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024428.1 | NP_001019599.1 | 3 - 4 | 267 | 67 | |
GenBank mRNA | BC095132.1 | - | 3 - 4 | 267 | 67 | |
BC165009.1 | - | 2 - 3 | 255 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
