Assay Details
Gene Symbol: tlk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024951.2 | NP_001020122.2 | 6 - 7 | 728 | 63 | |
XM_017356442.2 | XP_017211931.1 | 7 - 8 | 772 | 63 | ||
GenBank mRNA | AF265345.1 | - | 6 - 7 | 693 | 63 | |
BC071299.1 | - | 6 - 7 | 718 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
