Assay Details
| Gene Symbol: mcm6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025533.1 | NP_001020704.1 | 2 - 3 | 282 | 77 | |
| GenBank mRNA | AB973821.1 | - | 2 - 3 | 229 | 77 | |
| BC096872.1 | - | 2 - 3 | 282 | 77 | ||
More Information
Set Membership: |
|

mcm6l
Genomic Map