Assay Details
| Gene Symbol: nudt17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030103.1 | NP_001025274.1 | 2 - 3 | 180 | 85 | |
| XM_005158274.4 | XP_005158331.1 | 2 - 3 | 115 | 85 | ||
| XM_021466694.1 | XP_021322369.1 | 2 - 3 | 182 | 85 | ||
| GenBank mRNA | BC097186.1 | - | 2 - 3 | 180 | 85 | |
More Information
Set Membership: |
|

nudt17
Genomic Map