Assay Details
| Gene Symbol: apol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030138.1 | NP_001025309.1 | 2 - 3 | 111 | 78 | |
| GenBank mRNA | BC115193.1 | - | 2 - 3 | 101 | 78 | |
More Information
Set Membership: |
|

apol1
Genomic Map