Assay Details
| Gene Symbol: eef1db | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030147.2 | NP_001025318.1 | 4 - 5 | 354 | 82 | |
| NM_001161414.1 | NP_001154886.1 | 5 - 6 | 426 | 82 | ||
| XM_005160918.4 | XP_005160975.1 | 7 - 8 | 1315 | 82 | ||
| XM_005160919.4 | XP_005160976.1 | 6 - 7 | 1243 | 82 | ||
| XM_005160920.4 | XP_005160977.1 | 6 - 7 | 468 | 82 | ||
| XM_017352493.2 | XP_017207982.1 | 6 - 7 | 1260 | 82 | ||
| GenBank mRNA | BC081476.1 | - | 4 - 5 | 336 | 82 | |
| BC097116.1 | - | 4 - 5 | 336 | 82 | ||
| BC134997.1 | - | 5 - 6 | 409 | 82 | ||
| EH506100.1 | - | 4 - 5 | 365 | 82 | ||
More Information
Set Membership: |
|

eef1db
Genomic Map