Assay Details
| Gene Symbol: bco2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040312.1 | NP_001035402.1 | 1 - 2 | 88 | 97 | |
| GenBank mRNA | BC115259.1 | - | 1 - 2 | 88 | 97 | |
More Information
Set Membership: |
|

bco2b
Genomic Map