Assay Details
| Gene Symbol: limk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042691.2 | NP_001036156.1 | 1 - 2 | 85 | 76 | |
| GenBank mRNA | BC163263.1 | - | 1 - 2 | 85 | 76 | |
| DQ679229.1 | - | 1 - 2 | 85 | 76 | ||
More Information
Set Membership: |
|

limk1a
Genomic Map